Product Details

SNP ID
rs11557568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180302468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGTGGAAGGACAGCAGCTGCAG[C/T]GGAATCACGCTCAGGATGCCCTGGA
Phenotype
MIM: 603865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFPT2 PubMed Links

Gene Details

Gene
GFPT2
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005110.3 2014 Silent Mutation NP_005101.1
XM_006714942.2 2014 Intron XP_006715005.1
XM_017010100.1 2014 Silent Mutation XP_016865589.1

View Full Product Details