Product Details

SNP ID
rs7464896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23195328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTCTTTTGGTTTTGTTTTCCA[A/G]TCTGGAGAACTTTTTTCTTCTTTGC
Phenotype
MIM: 603611
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF10A PubMed Links

Gene Details

Gene
TNFRSF10A
Gene Name
TNF receptor superfamily member 10a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003844.3 Intron NP_003835.3

View Full Product Details