Product Details

SNP ID
rs6982684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143858020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGAGAGAAAGAAATAGGAGCCC[C/G]GTCTCAGGGTCCAGGGTGGCCCTCT
Phenotype
MIM: 607553
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EPPK1 PubMed Links

Gene Details

Gene
EPPK1
Gene Name
epiplakin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031308.3 15477 Silent Mutation ACC,ACG T5078T NP_112598.3
XM_017013889.1 15477 Silent Mutation ACC,ACG T5111T XP_016869378.1
XM_017013890.1 15477 Silent Mutation ACC,ACG T5078T XP_016869379.1
XM_017013891.1 15477 Silent Mutation ACC,ACG T4150T XP_016869380.1
XM_017013892.1 15477 Silent Mutation ACC,ACG T4044T XP_016869381.1

View Full Product Details