Product Details

SNP ID
rs7873594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:86220267 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATAGCATATTAGGAAACAGGATC[C/T]GGAAATGTATTTTAATATCTACACA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf153 PubMed Links

Gene Details

Gene
C9orf153
Gene Name
chromosome 9 open reading frame 153
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276366.1 1843 UTR 3 NP_001263295.1
NM_001276367.1 1843 UTR 3 NP_001263296.1
NM_001276368.1 1843 UTR 3 NP_001263297.1

View Full Product Details