Product Details

SNP ID
rs11616459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52502265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAACAAAATCAGGGCAACAGCG[A/T]TTTTCAGGGAAGAAAGGAGTTAACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TPTE2P3 PubMed Links

Gene Details

Gene
TPTE2P3
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details