Product Details

SNP ID
rs6934384
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:135292049 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGTCCAGAACTCTGGATTATTTT[C/T]AAGAAGTCAGAAATCTGTATTTCTT
Phenotype
MIM: 608894
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AHI1 PubMed Links

Gene Details

Gene
AHI1
Gene Name
Abelson helper integration site 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134830.1 Intron NP_001128302.1
NM_001134831.1 Intron NP_001128303.1
NM_001134832.1 Intron NP_001128304.1
NM_017651.4 Intron NP_060121.3
XM_011535910.2 Intron XP_011534212.1
XM_011535911.2 Intron XP_011534213.1
XM_017010977.1 Intron XP_016866466.1
XM_017010978.1 Intron XP_016866467.1
XM_017010979.1 Intron XP_016866468.1
XM_017010980.1 Intron XP_016866469.1
XM_017010981.1 Intron XP_016866470.1
XM_017010982.1 Intron XP_016866471.1
XM_017010983.1 Intron XP_016866472.1
XM_017010984.1 Intron XP_016866473.1

View Full Product Details