Product Details

SNP ID
rs8003994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:67821943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTCCCATGGAGAGAAATGAGGA[A/G]GATGTATTTTTGTTTGTGAGAGGTG
Phenotype
MIM: 602948 MIM: 612012
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51B PubMed Links

Gene Details

Gene
RAD51B
Gene Name
RAD51 paralog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321809.1 Intron NP_001308738.1
NM_001321810.1 Intron NP_001308739.1
NM_001321812.1 Intron NP_001308741.1
NM_001321814.1 Intron NP_001308743.1
NM_001321815.1 Intron NP_001308744.1
NM_001321817.1 Intron NP_001308746.1
NM_001321818.1 Intron NP_001308747.1
NM_001321819.1 Intron NP_001308748.1
NM_001321821.1 Intron NP_001308750.1
NM_002877.5 Intron NP_002868.1
NM_133509.3 Intron NP_598193.2
NM_133510.3 Intron NP_598194.1
XM_011537050.2 Intron XP_011535352.1
XM_011537051.2 Intron XP_011535353.1
XM_017021545.1 Intron XP_016877034.1
XM_017021546.1 Intron XP_016877035.1
XM_017021547.1 Intron XP_016877036.1
XM_017021548.1 Intron XP_016877037.1
Gene
ZFYVE26
Gene Name
zinc finger FYVE-type containing 26
There are no transcripts associated with this gene.

View Full Product Details