Product Details

SNP ID
rs11844699
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:106485859 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGGGCAGAACAACTAGGATGTC[C/T]GTTTATGTCAGTAGTGTCTGTCTTT
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
LINC00221
Gene Name
long intergenic non-protein coding RNA 221
There are no transcripts associated with this gene.

View Full Product Details