Product Details

SNP ID
rs6872685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154200232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGTGCCAGGGCAGGGATTATA[C/G]CAAGTCCCATGTGCCGCCTGGATGC
Phenotype
MIM: 608043
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GALNT10 PubMed Links

Gene Details

Gene
GALNT10
Gene Name
polypeptide N-acetylgalactosaminyltransferase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198321.3 Intron NP_938080.1

View Full Product Details