Product Details

SNP ID
rs7644811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:196354274 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTACTTAATCATCCCACAGTA[A/G]CCCGAGTTGAAGAAGTTACTTTGGT
Phenotype
MIM: 616379
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBXN7 PubMed Links

Gene Details

Gene
UBXN7
Gene Name
UBX domain protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015562.1 3904 UTR 3 NP_056377.1
XM_011512670.2 3904 UTR 3 XP_011510972.1
XM_011512671.2 3904 UTR 3 XP_011510973.1

View Full Product Details