Product Details

SNP ID
rs5744306
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:86470627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTCTCGCTACAATTATTAAATT[G/T]TCTTTCCAGTATGGATCCTAAAGTT
Phenotype
MIM: 603906
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLCA1 PubMed Links

Gene Details

Gene
CLCA1
Gene Name
chloride channel accessory 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285.3 Intron NP_001276.2

View Full Product Details