Product Details

SNP ID
rs17581532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55554347 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGAAGCACTCAATAATTTTGCAC[A/T]TGGATGTACCAACTTGTTAATGACT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4C15 PubMed Links

Gene Details

Gene
OR4C15
Gene Name
olfactory receptor family 4 subfamily C member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001920.1 41 Missense Mutation CAT,CTT H14L NP_001001920.1

View Full Product Details