Product Details

SNP ID
rs12783095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:45376762 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATTTTTTCCTTGATAAAGTGAG[A/T]CACAGGTCCCTTGTTATTCTCATCA
Phenotype
MIM: 152390
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ALOX5 PubMed Links

Gene Details

Gene
ALOX5
Gene Name
arachidonate 5-lipoxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000698.4 Intron NP_000689.1
NM_001256153.2 Intron NP_001243082.1
NM_001256154.2 Intron NP_001243083.1
NM_001320861.1 Intron NP_001307790.1
NM_001320862.1 Intron NP_001307791.1
XM_017016012.1 Intron XP_016871501.1

View Full Product Details