Product Details
- SNP ID
-
rs7075981
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124447543 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAGGCGGCGGACGGGAAGAGGAC[A/G]TTGGCCGCGGAGTAGGAGGGGAAAG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NKX1-2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs7089970] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NKX1-2
- Gene Name
- NK1 homeobox 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001146340.1 |
857 |
Silent Mutation |
AAC,AAT |
N273N |
NP_001139812.1 |
View Full Product Details