Product Details

SNP ID
rs11555893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:697007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCACCGGCGTGAGGCCTGCCCA[G/T]AACTTGGGCTGGTACAGCACGCGCC
Phenotype
MIM: 609082
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXL16 PubMed Links

Gene Details

Gene
FBXL16
Gene Name
F-box and leucine rich repeat protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153350.3 727 Missense Mutation TTA,TTC L133F NP_699181.2
Gene
WDR24
Gene Name
WD repeat domain 24
There are no transcripts associated with this gene.

View Full Product Details