Product Details

SNP ID
rs7913112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5862337 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTTGTGTTTCTTTCTTTCTGT[C/T]GGTGGTTCCTGAGGGTTGTGACGAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANKRD16 PubMed Links

Gene Details

Gene
ANKRD16
Gene Name
ankyrin repeat domain 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009941.2 2017 Intron NP_001009941.1
NM_001009943.2 2017 Intron NP_001009943.1
NM_019046.2 2017 UTR 3 NP_061919.1

View Full Product Details