Product Details

SNP ID
rs2477932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:26931835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGTCTGCCCCCTGGGGAGCCTCC[A/G]TCCCTCAGTCACTGAGACCCCTGGT
Phenotype
MIM: 610855
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD26 PubMed Links

Gene Details

Gene
ANKRD26
Gene Name
ankyrin repeat domain 26
There are no transcripts associated with this gene.

Gene
LINC00202-1
Gene Name
long intergenic non-protein coding RNA 202-1
There are no transcripts associated with this gene.

View Full Product Details