Product Details
- SNP ID
-
hCV57462825
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:1785791 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCCTCAGAATCCAACAGCAGTCG[C/T]TTGCAACAGAACTTTTTTTTTTTTA
- Phenotype
-
MIM: 139380
MIM: 611616
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GNB1
PubMed Links
Gene Details
- Gene
- GNB1
- Gene Name
- G protein subunit beta 1
There are no transcripts associated with this gene.
- Gene
- NADK
- Gene Name
- NAD kinase
There are no transcripts associated with this gene.
View Full Product Details