Product Details

SNP ID
rs28712052
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:1925510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGGGTGGGGGAGCTCTGTGCA[C/G]GGGCTGAGGAGGAGGGACAAAGAGC
Phenotype
MIM: 610171
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CALML6 PubMed Links

Gene Details

Gene
CALML6
Gene Name
calmodulin like 6
There are no transcripts associated with this gene.

Gene
CFAP74
Gene Name
cilia and flagella associated protein 74
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304360.1 Intron NP_001291289.1
XM_011542333.2 Intron XP_011540635.2
XM_011542335.1 Intron XP_011540637.1
XM_011542336.2 Intron XP_011540638.1
XM_017002641.1 Intron XP_016858130.1
XM_017002642.1 Intron XP_016858131.1
XM_017002643.1 Intron XP_016858132.1
XM_017002644.1 Intron XP_016858133.1
Gene
TMEM52
Gene Name
transmembrane protein 52
There are no transcripts associated with this gene.

View Full Product Details