Product Details

SNP ID
rs28709994
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:2051107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTTCTCCACGTTTACCCGGTCGG[A/G]GGTCTCGCTTTAGCCAAAATCCGGA
Phenotype
MIM: 176982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105378591 PubMed Links

Gene Details

Gene
LOC105378591
Gene Name
uncharacterized LOC105378591
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 Intron NP_001028753.1
NM_001033582.1 Intron NP_001028754.1
NM_001242874.1 Intron NP_001229803.1
NM_002744.4 Intron NP_002735.3
XM_011541773.1 Intron XP_011540075.1
XM_011541774.1 Intron XP_011540076.1
XM_011541775.2 Intron XP_011540077.1
XM_011541776.1 Intron XP_011540078.1
XM_011541778.1 Intron XP_011540080.1
XM_017001789.1 Intron XP_016857278.1
XM_017001790.1 Intron XP_016857279.1
XM_017001791.1 Intron XP_016857280.1
XM_017001792.1 Intron XP_016857281.1
XM_017001793.1 Intron XP_016857282.1
XM_017001794.1 Intron XP_016857283.1
XM_017001795.1 Intron XP_016857284.1
XM_017001796.1 Intron XP_016857285.1
XM_017001797.1 Intron XP_016857286.1
XM_017001798.1 Intron XP_016857287.1
XM_017001799.1 Intron XP_016857288.1
XM_017001800.1 Intron XP_016857289.1
XM_017001801.1 Intron XP_016857290.1
XM_017001802.1 Intron XP_016857291.1
XM_017001803.1 Intron XP_016857292.1
XM_017001804.1 Intron XP_016857293.1
XM_017001805.1 Intron XP_016857294.1

View Full Product Details