Product Details

SNP ID
rs34985495
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:161039757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTCCACATTGTGTGTTTCACACCA[C/T]TGCAGGCCGCCATATCACAGGGCCT
Phenotype
MIM: 614264 MIM: 616041 MIM: 191523
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
TSTD1
Gene Name
thiosulfate sulfurtransferase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113205.1 1303 Intron NP_001106676.1
NM_001113206.1 1303 Intron NP_001106677.1
NM_001113207.1 1303 Intron NP_001106678.1
Gene
USF1
Gene Name
upstream transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276373.1 1303 UTR 3 NP_001263302.1
NM_007122.4 1303 UTR 3 NP_009053.1
NM_207005.2 1303 UTR 3 NP_996888.1

View Full Product Details