Product Details

SNP ID
rs28584213
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:150226847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGACTGGGTAAATGTTTCTTCC[G/T]AGGATGTTGATGCTTTTTACCCTTG
Phenotype
MIM: 609611
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANP32E PubMed Links

Gene Details

Gene
ANP32E
Gene Name
acidic nuclear phosphoprotein 32 family member E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136478.3 Intron NP_001129950.1
NM_001136479.2 Intron NP_001129951.1
NM_001280559.1 Intron NP_001267488.1
NM_001280560.1 Intron NP_001267489.1
NM_030920.4 Intron NP_112182.1
XM_005245513.3 Intron XP_005245570.1
XM_005245514.3 Intron XP_005245571.1
XM_017002418.1 Intron XP_016857907.1

View Full Product Details