Product Details

SNP ID
rs28467468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184367058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGAGACTTGCTTCCCCAGGAA[C/T]AAATGCAGCATTAAGTGATTAGACT
Phenotype
MIM: 600570 MIM: 606023 MIM: 600044
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLCN2 PubMed Links

Gene Details

Gene
CLCN2
Gene Name
chloride voltage-gated channel 2
There are no transcripts associated with this gene.

Gene
POLR2H
Gene Name
RNA polymerase II subunit H
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278698.1 Intron NP_001265627.1
NM_001278699.1 Intron NP_001265628.1
NM_001278700.1 Intron NP_001265629.1
NM_001278714.1 Intron NP_001265643.1
NM_001278715.1 Intron NP_001265644.1
NM_006232.3 Intron NP_006223.2
XM_006713666.2 Intron XP_006713729.1
XM_006713667.2 Intron XP_006713730.1
XM_006713668.3 Intron XP_006713731.1
XM_006713670.2 Intron XP_006713733.1
XM_017006636.1 Intron XP_016862125.1
XM_017006637.1 Intron XP_016862126.1
Gene
THPO
Gene Name
thrombopoietin
There are no transcripts associated with this gene.

View Full Product Details