Product Details

SNP ID
rs28750322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47032301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGTCCCCTGAAAGGGGTGGTGG[G/T]GGGAGCAGGGAGGGAGCCCGTTAAG
Phenotype
MIM: 137190
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRB1 PubMed Links

Gene Details

Gene
GABRB1
Gene Name
gamma-aminobutyric acid type A receptor beta1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000812.3 Intron NP_000803.2
XM_017007985.1 Intron XP_016863474.1
XM_017007986.1 Intron XP_016863475.1

View Full Product Details