Product Details

SNP ID
rs28562785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69509542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCATGGCTGCCTTTTGTTTCATTG[C/T]CGCGTTGGTGATCTTTGTTACCAGT
Phenotype
MIM: 602876
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OCLN PubMed Links

Gene Details

Gene
OCLN
Gene Name
occludin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001205254.1 888 Missense Mutation GCC,GTC A151V NP_001192183.1
NM_001205255.1 888 Intron NP_001192184.1
NM_002538.3 888 Missense Mutation GCC,GTC A151V NP_002529.1
XM_017008913.1 888 Missense Mutation GCC,GTC A151V XP_016864402.1
XM_017008914.1 888 Missense Mutation GCC,GTC A151V XP_016864403.1

View Full Product Details