Product Details

SNP ID
rs28625719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:169791330 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTGCGATCATTAATTGTTCAC[A/G]TCTGTGAATACCCCCTGAGAGACAG
Phenotype
MIM: 615532
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMARD PubMed Links

Gene Details

Gene
ERMARD
Gene Name
ER membrane associated RNA degradation
There are no transcripts associated with this gene.

Gene
LINC00242
Gene Name
long intergenic non-protein coding RNA 242
There are no transcripts associated with this gene.

Gene
LINC00574
Gene Name
long intergenic non-protein coding RNA 574
There are no transcripts associated with this gene.

View Full Product Details