Product Details
- SNP ID
-
rs9347348
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:159795150 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTTCCCTAACTCAGTAGATGGAA[C/T]GTACAATCGGGTTTTATACCGAGAC
- Phenotype
-
MIM: 611831
MIM: 186980
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MRPL18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78069359] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MRPL18
- Gene Name
- mitochondrial ribosomal protein L18
- Gene
- PNLDC1
- Gene Name
- PARN like, ribonuclease domain containing 1
There are no transcripts associated with this gene.
- Gene
- SNORA29
- Gene Name
- small nucleolar RNA, H/ACA box 29
There are no transcripts associated with this gene.
- Gene
- TCP1
- Gene Name
- t-complex 1
There are no transcripts associated with this gene.
View Full Product Details