Product Details

SNP ID
rs11554013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36135777 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACCTGGTGATGCCCTTCATGCA[G/T]ACGGATCTGCAGAAGATCATGGGGA
Phenotype
MIM: 602899
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MAPK13 PubMed Links

Gene Details

Gene
MAPK13
Gene Name
mitogen-activated protein kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002754.4 432 Missense Mutation CAG,CAT Q111H NP_002745.1

View Full Product Details