Product Details

SNP ID
rs28364279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:87503991 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGAAAATATAAACAAAATTACACA[G/T]TTTATCTTTTAAAATCTACTTTAAT
Phenotype
MIM: 171050
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCB1 PubMed Links

Gene Details

Gene
ABCB1
Gene Name
ATP binding cassette subfamily B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000927.4 4588 UTR 3 NP_000918.2

View Full Product Details