Product Details

SNP ID
rs17876164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:95406665 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATTGAAAGAACTGACACTGTGC[A/T]TAAACCTTCATTTGAGAATTCATCT
Phenotype
MIM: 602447
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PON2 PubMed Links

Gene Details

Gene
PON2
Gene Name
paraoxonase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000305.2 Intron NP_000296.2
NM_001018161.1 Intron NP_001018171.1
XM_005250453.1 Intron XP_005250510.1
XM_017012357.1 Intron XP_016867846.1
XM_017012358.1 Intron XP_016867847.1

View Full Product Details