Product Details

SNP ID
rs28458449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109342449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTATACTTAAAATATATATTATG[C/T]ATGTTTATATAGTTTATGTATGTAT
Phenotype
MIM: 606109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENY2 PubMed Links

Gene Details

Gene
ENY2
Gene Name
ENY2, transcription and export complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193557.1 Intron NP_001180486.1
NM_020189.5 Intron NP_064574.1
Gene
NUDCD1
Gene Name
NudC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details