Product Details

SNP ID
rs28559999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34554893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGCCAGTGAGTCACAGAATGG[C/G]GCTAGGATGCACTACAGGCCACGCA
Phenotype
MIM: 118946
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNTFR PubMed Links

Gene Details

Gene
CNTFR
Gene Name
ciliary neurotrophic factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207011.1 Intron NP_001193940.1
NM_001842.4 Intron NP_001833.1
NM_147164.2 Intron NP_671693.1
XM_017014260.1 Intron XP_016869749.1
XM_017014261.1 Intron XP_016869750.1
XM_017014262.1 Intron XP_016869751.1
XM_017014263.1 Intron XP_016869752.1
XM_017014264.1 Intron XP_016869753.1
XM_017014265.1 Intron XP_016869754.1

View Full Product Details