Product Details
- SNP ID
-
rs17352063
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129008781 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGGTCCATGCCAGGCAGGCTGTAG[A/G]CAGTGATGAGCTGCAGAGATGGCAG
- Phenotype
-
MIM: 615587
MIM: 609288
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NUP188
PubMed Links
Gene Details
- Gene
- NUP188
- Gene Name
- nucleoporin 188
There are no transcripts associated with this gene.
- Gene
- SH3GLB2
- Gene Name
- SH3 domain containing GRB2 like endophilin B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001287045.1 |
1346 |
Missense Mutation |
GCC,GTC |
A373V |
NP_001273974.1 |
NM_001287046.1 |
1346 |
Missense Mutation |
GCC,GTC |
A364V |
NP_001273975.1 |
NM_020145.3 |
1346 |
Missense Mutation |
GCC,GTC |
A364V |
NP_064530.1 |
XM_005252098.2 |
1346 |
Missense Mutation |
GCC,GTC |
A369V |
XP_005252155.1 |
XM_005252100.2 |
1346 |
Missense Mutation |
GCC,GTC |
A348V |
XP_005252157.1 |
XM_005252101.3 |
1346 |
Missense Mutation |
GCC,GTC |
A94V |
XP_005252158.1 |
XM_006717188.2 |
1346 |
Missense Mutation |
GCC,GTC |
A397V |
XP_006717251.1 |
XM_006717189.2 |
1346 |
Missense Mutation |
GCC,GTC |
A393V |
XP_006717252.1 |
XM_006717190.1 |
1346 |
Missense Mutation |
GCC,GTC |
A392V |
XP_006717253.1 |
XM_006717191.1 |
1346 |
Missense Mutation |
GCC,GTC |
A368V |
XP_006717254.1 |
View Full Product Details