Product Details

SNP ID
rs28524858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:75402527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCCATTCTTACTCTCCCACCCT[C/G]TTTTCCTCCCTCCCCCCCTTCTTTG
Phenotype
MIM: 613902
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929234 PubMed Links

Gene Details

Gene
LOC101929234
Gene Name
uncharacterized LOC101929234
There are no transcripts associated with this gene.

Gene
ZNF503
Gene Name
zinc finger protein 503
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032772.5 Intron NP_116161.2
Gene
ZNF503-AS2
Gene Name
ZNF503 antisense RNA 2
There are no transcripts associated with this gene.

View Full Product Details