Product Details

SNP ID
rs35780811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:14968304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCTTACCTGGAAGAGGAGCGG[A/C]GTTTGCAGCATCAAGTCCTAGAGGT
Phenotype
MIM: 114130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CALCA PubMed Links

Gene Details

Gene
CALCA
Gene Name
calcitonin related polypeptide alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033952.2 1071 Intron NP_001029124.1
NM_001033953.2 1071 Intron NP_001029125.1
NM_001741.2 1071 Intron NP_001732.1
XM_017018283.1 1071 UTR 3 XP_016873772.1
XM_017018284.1 1071 UTR 3 XP_016873773.1

View Full Product Details