Product Details

SNP ID
rs11550521
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:17075177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCCGGCTCTCTATTAGAATCAG[A/G]CGGAATTTAGCATCCTTATCCTAAA
Phenotype
MIM: 180476
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPS13 PubMed Links

Gene Details

Gene
RPS13
Gene Name
ribosomal protein S13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017.2 Intron NP_001008.1
Gene
SNORD14A
Gene Name
small nucleolar RNA, C/D box 14A
There are no transcripts associated with this gene.

Gene
SNORD14B
Gene Name
small nucleolar RNA, C/D box 14B
There are no transcripts associated with this gene.

View Full Product Details