Product Details

SNP ID
rs28757209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:51210265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAATTGGGCTTAATTCACAGCAA[C/G]GGTCAAATGCTGAATTTCTAAGCAT
Phenotype
MIM: 107910
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYP19A1 PubMed Links

Gene Details

Gene
CYP19A1
Gene Name
cytochrome P450 family 19 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000103.3 2306 UTR 3 NP_000094.2
NM_031226.2 2306 UTR 3 NP_112503.1
XM_005254191.1 2306 UTR 3 XP_005254248.1
Gene
PIRC66
Gene Name
piwi-interacting RNA cluster 66
There are no transcripts associated with this gene.

View Full Product Details