Product Details

SNP ID
rs28675295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67824062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGGGGTACGCTCTCGGCTCGGG[A/C]GCTGTCTTCACTGTATGCGGGCCAG
Phenotype
MIM: 611273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SKOR1 PubMed Links

Gene Details

Gene
SKOR1
Gene Name
SKI family transcriptional corepressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258024.1 Intron NP_001244953.1
XM_017022173.1 Intron XP_016877662.1

View Full Product Details