Product Details

SNP ID
rs28710821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73928434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGTGCGCAGGAGGTTAGAGGGGA[A/G]CGTGTGCTCCAGGCAGGAGTCCGGA
Phenotype
MIM: 153456 MIM: 616800
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOXL1 PubMed Links

Gene Details

Gene
LOXL1
Gene Name
lysyl oxidase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005576.3 Intron NP_005567.2
XM_011521555.1 Intron XP_011519857.1
XM_017022179.1 Intron XP_016877668.1
Gene
LOXL1-AS1
Gene Name
LOXL1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details