Product Details

SNP ID
rs28418333
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:74614480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGCATGCGCCACCCAGTCCGGC[A/T]AATTTTTGTAATTTTTTGGAAGGGG
Phenotype
MIM: 602990
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CLK3 PubMed Links

Gene Details

Gene
CLK3
Gene Name
CDC like kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130028.1 Intron NP_001123500.1
NM_003992.4 Intron NP_003983.2
XM_005254151.3 Intron XP_005254208.1
XM_011521205.2 Intron XP_011519507.1
XM_011521206.2 Intron XP_011519508.1
XM_011521209.2 Intron XP_011519511.2
XM_017021905.1 Intron XP_016877394.1
XM_017021906.1 Intron XP_016877395.1
XM_017021907.1 Intron XP_016877396.1
XM_017021908.1 Intron XP_016877397.1
XM_017021909.1 Intron XP_016877398.1
XM_017021910.1 Intron XP_016877399.1
Gene
LOC102723750
Gene Name
uncharacterized LOC102723750
There are no transcripts associated with this gene.

View Full Product Details