Product Details
- SNP ID
-
rs28695985
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:24833873 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCATGTAGAATGAGGGCTCAGGA[A/C]ATATCATAATTTGAGAAGAAGAAAG
- Phenotype
-
MIM: 610238
MIM: 610739
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC5A11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75764559] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC5A11
- Gene Name
- solute carrier family 5 member 11
- Gene
- TNRC6A
- Gene Name
- trinucleotide repeat containing 6A
There are no transcripts associated with this gene.
View Full Product Details