Product Details

SNP ID
rs28650906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31394686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGAAATTCCTGATTATTTTTA[A/T]TTTTATTTTTACTCCAGCTCTGTTA
Phenotype
MIM: 602453
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ITGAD PubMed Links

Gene Details

Gene
ITGAD
Gene Name
integrin subunit alpha D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318185.1 Intron NP_001305114.1
NM_005353.2 Intron NP_005344.2
XM_011545835.2 Intron XP_011544137.1
XM_011545836.2 Intron XP_011544138.1
XM_011545837.2 Intron XP_011544139.1
XM_011545838.2 Intron XP_011544140.1
XM_011545839.2 Intron XP_011544141.1
XM_011545841.2 Intron XP_011544143.1
XM_011545842.2 Intron XP_011544144.1
XM_011545843.2 Intron XP_011544145.1
XM_011545844.2 Intron XP_011544146.1
XM_011545845.2 Intron XP_011544147.1
XM_011545846.2 Intron XP_011544148.1
XM_011545847.2 Intron XP_011544149.1
XM_011545848.2 Intron XP_011544150.1
XM_017023215.1 Intron XP_016878704.1

View Full Product Details