Product Details

SNP ID
rs28756438
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67338850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAAGCTTTTCAAATTTCTAATTT[C/T]TGCTTGGAAGCTGAATTTTTTTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC36 PubMed Links

Gene Details

Gene
LRRC36
Gene Name
leucine rich repeat containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161575.1 Intron NP_001155047.1
NM_018296.5 Intron NP_060766.5
XM_005256025.2 Intron XP_005256082.1
XM_005256026.2 Intron XP_005256083.1
XM_005256027.2 Intron XP_005256084.1
XM_005256028.1 Intron XP_005256085.1
XM_011523200.1 Intron XP_011521502.1
XM_011523201.1 Intron XP_011521503.1
XM_011523202.2 Intron XP_011521504.1
XM_011523203.1 Intron XP_011521505.1
XM_011523204.1 Intron XP_011521506.1
XM_011523205.1 Intron XP_011521507.1
XM_011523206.2 Intron XP_011521508.1
XM_017023400.1 Intron XP_016878889.1
XM_017023401.1 Intron XP_016878890.1
XM_017023402.1 Intron XP_016878891.1

View Full Product Details