Product Details
- SNP ID
-
rs28715141
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69204413 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTCTTATTCCATATATGGAATAT[A/T]ATTGCTCCTCAAAAGAAAAACATGT
- Phenotype
-
MIM: 600027
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNTB2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76178191] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNTB2
- Gene Name
- syntrophin beta 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006750.3 |
|
Intron |
|
|
NP_006741.1 |
View Full Product Details