Product Details

SNP ID
rs28575510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:86557033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTACTCCACTTGGGCGACACAGC[C/G]AGAACTCCTCTTTGAAAAACATAAT
Phenotype
MIM: 616820
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FLJ30679 PubMed Links

Gene Details

Gene
FLJ30679
Gene Name
uncharacterized protein FLJ30679
There are no transcripts associated with this gene.

Gene
FOXC2-AS1
Gene Name
FOXC2 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MTHFSD
Gene Name
methenyltetrahydrofolate synthetase domain containing
There are no transcripts associated with this gene.

View Full Product Details