Product Details

SNP ID
rs28632046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78382332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGTATGATTTCTTTTGATTCT[A/G]GTTCAGCCTGAGATGGTGGGGGCTT
Phenotype
MIM: 614942
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928674 PubMed Links

Gene Details

Gene
LOC101928674
Gene Name
uncharacterized LOC101928674
There are no transcripts associated with this gene.

Gene
PGS1
Gene Name
phosphatidylglycerophosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024419.4 Intron NP_077733.3
XM_011525487.2 Intron XP_011523789.1
XM_011525488.2 Intron XP_011523790.1
XM_011525489.2 Intron XP_011523791.1
XM_017025357.1 Intron XP_016880846.1
XM_017025358.1 Intron XP_016880847.1
XM_017025359.1 Intron XP_016880848.1
XM_017025360.1 Intron XP_016880849.1
XM_017025361.1 Intron XP_016880850.1
XM_017025362.1 Intron XP_016880851.1
XM_017025363.1 Intron XP_016880852.1
XM_017025364.1 Intron XP_016880853.1
XM_017025365.1 Intron XP_016880854.1
XM_017025366.1 Intron XP_016880855.1

View Full Product Details