Product Details

SNP ID
rs28750125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15976778 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGTCCACTGCTTCAACTCCTT[C/T]TGCAGTATCCCTGCAACAATATTAC
Phenotype
MIM: 600446 MIM: 614535
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA2B PubMed Links

Gene Details

Gene
ADORA2B
Gene Name
adenosine A2b receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000676.2 1660 Intron NP_000667.1
XM_011523659.2 1660 Intron XP_011521961.1
XM_011523661.2 1660 Intron XP_011521963.1
XM_017024197.1 1660 Intron XP_016879686.1
Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 1660 UTR 3 NP_001036162.1
NM_001042698.1 1660 UTR 3 NP_001036163.1

View Full Product Details