Product Details

SNP ID
rs8078543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68041135 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGAGACATGCATTTAGAATGATA[G/T]ATCCATTTAGGAGATAAGTATAATT
Phenotype
MIM: 600685
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KPNA2 PubMed Links

Gene Details

Gene
KPNA2
Gene Name
karyopherin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320611.1 Intron NP_001307540.1
NM_002266.3 Intron NP_002257.1

View Full Product Details