Product Details
- SNP ID
-
rs11541800
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:31592953 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAGTTCTAGATGCTGTCCGAGGC[A/G]GTCCTGCCATCAATGTGGCCGTGCA
- Phenotype
-
MIM: 176300
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TTR
PubMed Links
Gene Details
- Gene
- TTR
- Gene Name
- transthyretin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000371.3 |
263 |
Missense Mutation |
AGT,GGT |
S43G |
NP_000362.1 |
View Full Product Details