Product Details

SNP ID
rs34014819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45406376 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTGTGGCTGCCCCTTTAAGGAG[G/T]CGTTGCAACAAACCATATTGGACAG
Phenotype
MIM: 107325 MIM: 126380 MIM: 607463
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CD3EAP PubMed Links

Gene Details

Gene
CD3EAP
Gene Name
CD3e molecule associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297590.1 248 UTR 5 NP_001284519.1
NM_012099.1 248 UTR 5 NP_036231.1
XM_017026180.1 248 UTR 5 XP_016881669.1
XM_017026181.1 248 UTR 5 XP_016881670.1
Gene
ERCC1
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166049.1 248 Intron NP_001159521.1
NM_001983.3 248 Intron NP_001974.1
NM_202001.2 248 Intron NP_973730.1
XM_005258634.1 248 Intron XP_005258691.1
XM_005258635.2 248 Intron XP_005258692.1
XM_005258636.4 248 Intron XP_005258693.1
XM_005258637.1 248 Intron XP_005258694.1
XM_011526610.2 248 Intron XP_011524912.1
XM_017026459.1 248 Intron XP_016881948.1
XM_017026460.1 248 Intron XP_016881949.1
XM_017026461.1 248 Intron XP_016881950.1
XM_017026462.1 248 Intron XP_016881951.1
XM_017026463.1 248 Intron XP_016881952.1
XM_017026464.1 248 Intron XP_016881953.1
XM_017026465.1 248 Intron XP_016881954.1
XM_017026466.1 248 Intron XP_016881955.1
Gene
PPP1R13L
Gene Name
protein phosphatase 1 regulatory subunit 13 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142502.1 248 Intron NP_001135974.1
NM_006663.3 248 Intron NP_006654.2
XM_017026177.1 248 Intron XP_016881666.1
XM_017026178.1 248 Intron XP_016881667.1
XM_017026179.1 248 Intron XP_016881668.1

View Full Product Details